Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172635.3 | NP_766223.2 | 8 - 9 | 1199 | 65 | |
GenBank mRNA | AK085013.1 | - | 8 - 9 | 1141 | 65 | |
AK087559.1 | - | 8 - 9 | 1163 | 65 | ||
AK150301.1 | - | 8 - 9 | 1200 | 65 | ||
AK154102.1 | - | 8 - 9 | 1197 | 65 | ||
AK154873.1 | - | 8 - 9 | 1151 | 65 | ||
AK161738.1 | - | 1 - 2 | 160 | 65 | ||
AK170408.1 | - | 8 - 9 | 1163 | 65 | ||
AK170745.1 | - | 8 - 9 | 1164 | 65 | ||
AK170918.1 | - | 8 - 9 | 1200 | 65 | ||
BC047458.1 | - | 8 - 9 | 1001 | 65 | ||
BC058941.1 | - | 8 - 9 | 1140 | 6825758 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |