High-performance secondary antibodies

Shop now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm01203005_m1
            See other SGMS1 GE Assays ›
            Gene Symbol:
            Sgms1
            Gene Name:
            sphingomyelin synthase 1
            Gene Aliases:
            9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
            Chromosome Location:
            Chr.19: 32122727 - 32389705 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm01203005_m1
            Size
            Availability Made To Order
            Catalog # 4351372
            Price (USD) 417.00
            Your Price
            Online offer (USD):387.65
            387.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: Sgms1
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_144792.4 NP_659041.3 3 - 4 405 92
            GenBank mRNA AK031448.1 - 1 - 2 40 92
            AK082974.1 - 3 - 4 334 92
            AK146782.1 - 3 - 4 405 92
            AK151170.1 - 3 - 4 390 92
            AK153406.1 - 3 - 4 390 92
            AY509044.1 - 3 - 4 405 92

            Back To Top

            More Information


            Additional Information:

            This assay is in the 5'UTR region of the AK082974.1, AK146782.1, AK151170.1, AK153406.1, AY509044.1, NM_144792.4 transcripts.

            Set Membership:

            Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene Sgms1

            Gene Ontology Categories:

            Function(s) Process(es)

            lipid metabolic process
            sphingolipid metabolic process
            sphingomyelin biosynthetic process
            apoptotic process
            inflammatory response
            positive regulation of gene expression
            cell growth
            phosphorylation
            negative regulation of extrinsic apoptotic signaling pathway
            kinase activity
            transferase activity
            sphingomyelin synthase activity
            ceramide cholinephosphotransferase activity

            MicroArray Probes:

            Affymetrix ABI

            Array Name Probe IDs Targets Target Type
            Affymetrix Mouse Clariom S Array PSR1900012209.mm.1 NM_001168525.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012209.mm.1 NM_001168525.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012209.mm.1 NM_144792.4.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012209.mm.1 NM_144792.4 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012209.mm.1 TC1900001390.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR1900012221.mm.1 NM_001168525.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012221.mm.1 NM_001168525.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012221.mm.1 NM_144792.4.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012221.mm.1 NM_144792.4 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012221.mm.1 TC1900001390.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR1900012213.mm.1 NM_001168525.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012213.mm.1 NM_001168525.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012213.mm.1 NM_144792.4.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012213.mm.1 NM_144792.4 Refseq
            Affymetrix Mouse Clariom S Array PSR1900012213.mm.1 TC1900001390.mm.1 MTA TC
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 382962 - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-6cgcf:80/100.66.79.97:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline