Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291205.1 | NP_001278134.1 | 4 - 5 | 403 | 78 | |
NM_001291206.1 | NP_001278135.1 | 4 - 5 | 754 | 78 | ||
NM_001291207.1 | NP_001278136.1 | 3 - 4 | 299 | 78 | ||
NM_001291208.1 | NP_001278137.1 | 3 - 4 | 648 | 78 | ||
NM_001291209.1 | NP_001278138.1 | 4 - 5 | 405 | 78 | ||
NM_001291210.1 | NP_001278139.1 | 2 - 3 | 177 | 78 | ||
NM_026446.4 | NP_080722.1 | 4 - 5 | 405 | 78 | ||
GenBank mRNA | AF535869.1 | - | 4 - 5 | 755 | 78 | |
AF535870.1 | - | 3 - 4 | 649 | 78 | ||
AK011740.2 | - | 4 - 5 | 354 | 78 | ||
AK019401.1 | - | 4 - 5 | 378 | 78 | ||
AK042531.1 | - | 4 - 5 | 353 | 78 | ||
AK154844.1 | - | 4 - 5 | 405 | 78 | ||
AY138505.1 | - | 3 - 4 | 251 | 78 | ||
BC003838.1 | - | 4 - 5 | 337 | 3490927 | 78 | |
BG085760.2 | - | 4 - 5 | 556 | 78 | ||
BU694737.1 | - | 2 - 3 | 213 | 78 | ||
CF751080.1 | - | 3 - 4 | 319 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |