Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbm46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277170.1 | NP_001264099.1 | 2 - 3 | 335 | 146 | |
GenBank mRNA | CJ047211.1 | - | 2 - 3 | 336 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Probe spans exons |