Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145155.3 | NP_660137.1 | 5 - 6 | 854 | 61 | |
GenBank mRNA | AF454703.1 | - | 4 - 5 | 473 | 61 | |
AK044481.1 | - | 875 | 61 | |||
BC027038.1 | - | 5 - 6 | 854 | 5363785 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |