Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10588109 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0900009758.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009758.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009758.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009754.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009754.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009754.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009755.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009755.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009755.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009756.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009756.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009756.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009757.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009757.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009757.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009753.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009753.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009753.mm.1 |
TC0900001251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900009752.mm.1 |
NM_001165957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009752.mm.1 |
NM_001165957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900009752.mm.1 |
TC0900001251.mm.1 |
MTA TC |