Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100182.2 | NP_001093652.1 | 1 - 2 | 319 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Mus musculus Inventoried Probe spans exons |