Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900057B20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045365.1 | 1 - 2 | 253 | 112 | ||
GenBank mRNA | AK013712.1 | - | 1 - 2 | 256 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |