Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00019955.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019955.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019955.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019955.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019955.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019966.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019966.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019966.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019966.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019966.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019967.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019967.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019967.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019964.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019964.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019964.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019964.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019964.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019953.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019953.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019953.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019953.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019953.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019961.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019961.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019961.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019961.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019961.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019956.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019956.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019956.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019956.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019956.mm.1 |
TC0X00003173.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019954.mm.1 |
NM_001168600.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019954.mm.1 |
NM_001168600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019954.mm.1 |
NM_198110.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019954.mm.1 |
NM_198110.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019954.mm.1 |
TC0X00003173.mm.1 |
MTA TC |