Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001182.3 | NP_001001182.2 | 5 - 6 | 686 | 69 | |
GenBank mRNA | AK030867.1 | - | 4 - 5 | 602 | 69 | |
AK134250.1 | - | 6 - 7 | 765 | 69 | ||
AK147737.1 | - | 6 - 7 | 878 | 69 | ||
AK173180.1 | - | 6 - 7 | 784 | 69 | ||
BC027336.1 | - | 2 - 3 | 311 | 69 | ||
BC150814.1 | - | 6 - 7 | 668 | 9087725 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |