Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007459.3 | NP_031485.3 | 6 - 7 | 886 | 82 | |
GenBank mRNA | AK088500.1 | - | 6 - 7 | 820 | 82 | |
AK128972.1 | - | 6 - 7 | 829 | 82 | ||
AK152463.1 | - | 6 - 7 | 819 | 82 | ||
AK173054.1 | - | 6 - 7 | 823 | 82 | ||
BC010597.1 | - | 3 - 4 | 398 | 82 | ||
BC058099.1 | - | 6 - 7 | 828 | 6825406 | 82 | |
X14972.1 | - | 6 - 7 | 856 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |