Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039512.1 | NP_001034601.1 | 10 - 11 | 1620 | 79 | |
NM_054102.2 | NP_473443.2 | 11 - 12 | 1746 | 79 | ||
GenBank mRNA | AB055737.1 | - | 11 - 12 | 1742 | 79 | |
AK129229.1 | - | 11 - 12 | 1684 | 79 | ||
AK144619.1 | - | 11 - 12 | 1726 | 79 | ||
AK144835.1 | - | 10 - 11 | 1601 | 79 | ||
AK145071.1 | - | 11 - 12 | 1737 | 79 | ||
AK145845.1 | - | 11 - 12 | 1696 | 79 | ||
AK146001.1 | - | 11 - 12 | 1727 | 79 | ||
AK146329.1 | - | 7 - 8 | 944 | 79 | ||
AK146494.1 | - | 11 - 12 | 1699 | 79 | ||
AK146645.1 | - | 11 - 12 | 1697 | 79 | ||
AK159254.1 | - | 11 - 12 | 1728 | 79 | ||
BC004092.1 | - | 10 - 11 | 1446 | 3593827 | 79 | |
DQ914522.1 | - | 11 - 12 | 1578 | 79 | ||
DQ914523.1 | - | 11 - 12 | 2629 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |