Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB004817.1 | - | 14 - 15 | 1650 | 74 | |
AF483490.1 | - | 12 - 13 | 1539 | 74 | ||
AF483491.1 | - | 12 - 13 | 1539 | 74 | ||
AK003058.1 | - | 2 - 3 | 72 | 74 | ||
AK045374.1 | - | 14 - 15 | 1638 | 74 | ||
AK129004.1 | - | 13 - 14 | 1610 | 74 | ||
AK138942.1 | - | 14 - 15 | 1633 | 74 | ||
AK140694.1 | - | 14 - 15 | 1657 | 74 | ||
AK154565.1 | - | 14 - 15 | 1630 | 74 | ||
AK164192.1 | - | 14 - 15 | 1649 | 74 | ||
AK211176.1 | - | 1 - 2 | 45 | 74 | ||
BC007135.1 | - | 13 - 14 | 1610 | 3600884 | 74 | |
U60453.1 | - | 14 - 15 | 1638 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |