Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019637.3 | NP_062611.2 | 1 - 2 | 340 | 86 | |
GenBank mRNA | AK002822.1 | - | 282 | 86 | ||
AK030122.1 | - | 1 - 2 | 340 | 86 | ||
AK046970.1 | - | 1 - 2 | 317 | 86 | ||
AK166043.1 | - | 1 - 2 | 323 | 86 | ||
BC038608.1 | - | 1 - 2 | 290 | 5012303 | 86 | |
BC085089.1 | - | 1 - 2 | 287 | 6492329 | 86 | |
BC096674.1 | - | 1 - 2 | 348 | 5144003 | 86 | |
U34973.1 | - | 1 - 2 | 308 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |