Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026231.2 | NP_080507.2 | 4 - 5 | 1330 | 64 | |
GenBank mRNA | AK013967.1 | - | 1 - 2 | 182 | 64 | |
AK017059.1 | - | 2 - 3 | 605 | 64 | ||
AK020021.1 | - | 3 - 4 | 940 | 64 | ||
AK029724.1 | - | 4 - 5 | 1329 | 64 | ||
AK033409.1 | - | 4 - 5 | 1368 | 64 | ||
AK044441.1 | - | 4 - 5 | 1338 | 64 | ||
BC047136.1 | - | 4 - 5 | 1330 | 6311866 | 64 | |
BC061240.1 | - | 4 - 5 | 1326 | 6743615 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |