Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199015.4 | NP_950180.3 | 5 - 6 | 535 | 96 | |
GenBank mRNA | AK156583.1 | - | 5 - 6 | 516 | 96 | |
BC030186.1 | - | 5 - 6 | 536 | 5067268 | 96 | |
BC110663.1 | - | 4 - 5 | 1619 | 96 | ||
BC145673.1 | - | 5 - 6 | 455 | 40130976 | 96 | |
EU155107.1 | - | 2 - 3 | 146 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |