Boost your research budget through the Funding Plus program

Explore now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm01322694_m1
            See other 4930425O10RIK GE Assays ›
            Gene Symbol:
            4930425O10Rik
            Gene Name:
            RIKEN cDNA 4930425O10 gene
            Gene Aliases:
            -
            Chromosome Location:
            Chr.3: 138711535 - 138726984 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm01322694_m1
            Size
            Availability Made To Order
            Catalog # 4426961
            Price (USD) 416.00
            Your Price
            Online offer (USD):390.65
            390.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Gene Symbol: 4930425O10Rik
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NR_040545.1 3 - 4 275 70
            GenBank mRNA AK015203.1 - 3 - 4 275 70
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
            GenBankmRNA CD771906.1 - 153 Chr. - - [-] on Build 70
            GenBankmRNA BY714758.1 - 3 - 4 275 Chr. 3 - 138712293 - 138726984 [-] on Build GRCm38 70

            Adjacent Gene Details

            Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
            Upstream eukaryotic translation initiation factor 4E Eif4e EG668879,
            Eif4e-ps,
            If4e,
            eIF-4E
            -153.9kb Anti-sense Chr. 3 - 138526191 - 138557599 on NCBI Build GRCm38
            Downstream tetraspanin 5 Tspan5 2810455A09Rik,
            4930505M03Rik,
            NET-4,
            Tm4sf9
            27.2kb Anti-sense Chr. 3 - 138742208 - 138904433 on NCBI Build GRCm38

            Back To Top

            More Information


            Updated Mapping Information

            This assay targets the same location as MTA transcript cluster(s) TC0300003021.mm.1

            Set Membership:

            Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

            MicroArray Probes:

            Agilent Affymetrix ABI

            Array Name Probe IDs Targets Target Type
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2384248 AK015203.1 mouse_transcriptome
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 247688 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193653 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33887 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399309 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252924 - -
            Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501398 - -
            Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172110 - -
            Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177444 - -
            Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177444 - -
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112129 - -
            Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177444 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570237 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570262 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570340 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570235 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570298 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570308 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570349 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570204 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570312 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570335 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P03098536 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570352 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570201 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570279 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570345 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570294 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570319 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570311 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570347 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570302 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570211 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P03167181 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570343 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570357 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02651530 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02812870 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P03169997 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570315 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570310 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570333 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570348 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570328 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570266 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570232 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570297 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570309 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570358 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570231 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570203 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570275 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570282 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570281 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02781905 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570261 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570239 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570241 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570259 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570267 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570206 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02793457 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570215 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570233 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570323 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570277 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570287 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570321 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570256 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570300 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570236 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570336 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570286 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570325 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570305 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570289 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570208 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P03113315 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570199 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570230 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570304 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570292 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P03181506 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570270 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02479825 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570316 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570350 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570290 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570205 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570338 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570273 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02459403 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570353 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P02884572 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570212 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570257 - -
            Agilent Six_ChIP_01_of_03_v2 A_68_P22570210 - -
            Array Name Probe IDs Targets Target Type
            Affymetrix HT MG-430 PM Array Plate 1432085_PM_at - -
            Mouse Genome 430 2.0 Array 1432085_at - -
            Mouse Genome 430 2.0 Array 1432085_PM_at - -
            Affymetrix Mouse Clariom S Array PSR0300023853.mm.1 NR_040545.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR0300023853.mm.1 NR_040545.1 Refseq
            Affymetrix Mouse Clariom S Array PSR0300023853.mm.1 TC0300003021.mm.1 MTA TC
            Mouse Expression Set 430 1432085_at - -
            Mouse Expression Set 430 1432085_PM_at - -
            Affymetrix HT Mouse Genome 430B Array 1432085_PM_at - -
            Affymetrix HT Mouse Genome 430B Array 1432085_at - -
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 462275 - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-tw7ck:80/100.66.77.154:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline