Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029987.1 | NP_001025158.1 | 2 - 3 | 1843 | 88 | |
GenBank mRNA | AK144307.1 | - | 1 - 2 | 540 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |