Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026512.1 | NP_080788.1 | 4 - 5 | 550 | 75 | |
GenBank mRNA | AK008202.1 | - | 4 - 5 | 569 | 75 | |
AK017801.1 | - | 4 - 5 | 553 | 75 | ||
AK078232.1 | - | 4 - 5 | 595 | 75 | ||
AK152712.1 | - | 4 - 5 | 560 | 75 | ||
AK170099.1 | - | 4 - 5 | 553 | 75 | ||
BC023146.1 | - | 4 - 5 | 535 | 3990118 | 75 | |
BC025162.1 | - | 4 - 5 | 550 | 4921451 | 75 | |
BC029322.1 | - | 2 - 3 | 185 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |