Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081059.3 | NP_001074528.1 | 2 - 3 | 540 | 74 | |
GenBank mRNA | AK018089.1 | - | 2 - 3 | 511 | 74 | |
AK041849.1 | - | 2 - 3 | 541 | 74 | ||
AK210508.1 | - | 1 - 2 | 49 | 74 | ||
AK216207.1 | - | 1 - 2 | 49 | 74 | ||
AK219679.1 | - | 1 - 2 | 49 | 74 | ||
BC113195.1 | - | 3 - 4 | 278 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |