Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK053178.1 | - | 7 - 8 | 2617 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons |