Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm01722035_s1
            See other 2310034G01RIK GE Assays ›
            Gene Symbol:
            2310034G01Rik
            Gene Name:
            RIKEN cDNA 2310034G01 gene
            Gene Aliases:
            -
            Chromosome Location:
            Chr.19: 46348201 - 46348988 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm01722035_s1
            Size
            Availability Made To Order
            Catalog # 4426961
            Price (USD) 416.00
            Your Price
            Online offer (USD):390.65
            390.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: 2310034G01Rik
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NR_040418.1 1 428 83
            GenBank mRNA AK009611.1 - 1 395 83
            AK051784.1 - 1 415 83
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
            GenBankmRNA AW988748.1 - 3 159 Chr. 19 - 46348207 - 46350270 [-] on Build GRCm38 83
            GenBankmRNA BB463385.2 - 1 427 Chr. 19 - 46348518 - 46348978 [-] on Build GRCm38 83
            GenBankmRNA BB657258.1 - 1 415 Chr. 19 - 46348505 - 46348974 [-] on Build GRCm38 83
            GenBankmRNA BX529202.1 - 1 333 Chr. 19 - 46348347 - 46348871 [-] on Build GRCm38 83
            GenBankmRNA AI854627.1 - 1 Chr. 19 - 46348201 - 46348677 [+] on Build GRCm38 83
            GenBankmRNA W97823.1 - 287 Chr. - - [-] on Build 83
            GenBankmRNA DT904655.1 - 1 287 Chr. 19 - 46348444 - 46348839 [-] on Build GRCm38 83
            GenBankmRNA AI429807.1 - 1 Chr. 19 - 46348201 - 46348638 [+] on Build GRCm38 83
            GenBankmRNA BE863644.1 - 1 410 Chr. 19 - 46348480 - 46348970 [-] on Build GRCm38 83

            Adjacent Gene Details

            Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
            Upstream microRNA 146b Mir146b Mirn146b,
            mir-146b
            -5.5kb Anti-sense Chr. 19 - 46342762 - 46342870 on NCBI Build GRCm38
            Downstream transmembrane protein 180 Tmem180 4930449A08Rik,
            4930538D17Rik
            8.0kb Anti-sense Chr. 19 - 46356877 - 46375254 on NCBI Build GRCm38

            Back To Top

            More Information


            Additional Information:

            Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

            Set Membership:

            Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

            MicroArray Probes:

            Agilent Affymetrix ABI Illumina

            Array Name Probe IDs Targets Target Type
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118849 AI854627.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118849 AK009611.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123291 AI854627.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123291 AK009611.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123291 AK051784.1 mouse_transcriptome
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118849 AI854627.1 mouse_transcriptome
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118849 AK009611.1 mouse_transcriptome
            Array Name Probe IDs Targets Target Type
            Affymetrix HT MG-430 PM Array Plate 1440229_PM_at - -
            Mouse Genome 430 2.0 Array 1440229_PM_at - -
            Mouse Genome 430 2.0 Array 1440229_at - -
            Affymetrix GeneChip Mouse Genome 430 2.0 Array 75579_PM_at - -
            Affymetrix GeneChip Mouse Genome 430 2.0 Array 75579_at - -
            Affymetrix Mouse Clariom S Array PSR1900014262.mm.1 NR_040418.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900014262.mm.1 NR_040418.1 Refseq
            Affymetrix Mouse Clariom S Array PSR1900014262.mm.1 TC1900001601.mm.1 MTA TC
            Mouse Expression Set 430 1440229_at - -
            Mouse Expression Set 430 1440229_PM_at - -
            Affymetrix HT Mouse Genome 430B Array 1440229_at - -
            Affymetrix HT Mouse Genome 430B Array 1440229_PM_at - -
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 532954 - -
            Array Name Probe IDs Targets Target Type
            MouseWG-6 v2 Expression BeadChip 104230300 - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-v5vpt:80/100.66.77.154:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline