Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005131.2 | - | 5 | 937 | 70 | |
AK140877.1 | - | 5 | 991 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |