Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163713.1 | NP_001157185.1 | 6 - 7 | 902 | 66 | |
NM_172745.3 | NP_766333.1 | 6 - 7 | 902 | 66 | ||
GenBank mRNA | AK075857.1 | - | 6 - 7 | 826 | 66 | |
AK084724.1 | - | 6 - 7 | 883 | 66 | ||
AK152858.1 | - | 6 - 7 | 897 | 66 | ||
AK153135.1 | - | 6 - 7 | 897 | 66 | ||
BC060959.1 | - | 6 - 7 | 879 | 30379757 | 66 | |
BC100596.1 | - | 6 - 7 | 895 | 6762419 | 66 | |
BU699735.1 | - | 1 - 2 | 145 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |