Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052407.1 | - | 1446 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | S-adenosylhomocysteine hydrolase-like 2 | Ahcyl2 |
4631427C17Rik, AI227036, Adohcyase3, mKIAA0828 |
5.9kb | Sense | Chr. 6 - 29768379 - 29912310 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() |