Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pilra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153510.3 | NP_705730.1 | 3 - 4 | 828 | 77 | |
GenBank mRNA | AJ400844.1 | - | 726 | 77 | ||
AK041413.1 | - | 2 - 3 | 1119 | 77 | ||
AK041670.1 | - | 3 - 4 | 740 | 77 | ||
AK170000.1 | - | 3 - 4 | 740 | 77 | ||
BC115565.2 | - | 713 | 40102015 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |