Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177593.1 | NP_808261.1 | 3 - 4 | 424 | 96 | |
GenBank mRNA | BC037749.1 | - | 3 - 4 | 424 | 1429110 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |