Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255991.1 | NP_001242920.1 | 8 - 9 | 1018 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus Probe spans exons |