Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198166.3 | NP_937809.1 | 1 - 2 | 210 | 71 | |
GenBank mRNA | AB116020.1 | - | 1 - 2 | 157 | 71 | |
AK132025.1 | - | 1 - 2 | 211 | 71 | ||
AV144685.2 | - | 1 - 2 | 224 | 71 | ||
AY321315.1 | - | 1 - 2 | 191 | 71 | ||
BB611261.1 | - | 1 - 2 | 211 | 71 | ||
BC117024.1 | - | 1 - 2 | 182 | 40126343 | 71 | |
BC117026.1 | - | 1 - 2 | 182 | 40126345 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |