Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10441640 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441634 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441638 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441639 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441641 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441635 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441636 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10441637 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1450357_PM_a_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10441633 |
- |
- |
Mouse Genome 430 2.0 Array |
1450357_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1450357_a_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
12458_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
12458_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700000651.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000651.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000651.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_001190335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000652.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_001190338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000650.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000644.mm.1 |
NM_001190333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000644.mm.1 |
NM_001190333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000644.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190337.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190337.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_001190338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000653.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000645.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000645.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000645.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190337.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_001190337.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000648.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190333.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190333.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190334.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190334.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190337.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190337.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_001190338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_009835.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
NM_009835.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000655.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000649.mm.1 |
NM_001190338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000649.mm.1 |
NM_001190338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000649.mm.1 |
TC1700000084.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1450357_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1450357_a_at |
- |
- |
Murine Genome U74v2 Set |
99899_PM_at |
- |
- |
Murine Genome U74v2 Set |
99899_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450357_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450357_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1450357_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1450357_PM_a_at |
- |
- |