Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enhanced GFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U55761.1 | - | 418 | 68 |
Gene Symbol: YFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY189981_YFP.0 | - | 1924 | 68 |
Gene Symbol: YFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY189981_YFP.0 | - | 1363 | 68 |
Set Membership: |
![]() ![]() ![]() |