Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CX3CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082134.1 | NP_001075603.1 | 1 | 847 | 102 | |
XM_008260200.1 | XP_008258422.1 | 2 | 959 | 102 | ||
GenBank mRNA | AM183335.1 | - | 1 | 847 | 102 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |