Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032391.1 | 38 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
![]() ![]() ![]() |