Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021655.2 | NP_067687.2 | 7 - 8 | 1463 | 88 | |
GenBank mRNA | AF145445.1 | - | 7 - 8 | 1316 | 88 | |
BC087703.1 | - | 7 - 8 | 1443 | 7313842 | 88 | |
X06832.1 | - | 7 - 8 | 1451 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |