Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031513.1 | NP_113701.1 | 3 - 4 | 331 | 77 | |
XM_006246226.2 | XP_006246288.1 | 3 - 4 | 307 | 77 | ||
GenBank mRNA | U81492.1 | - | 3 - 4 | 331 | 77 | |
U81493.1 | - | 3 - 4 | 322 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |