Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031641.1 | NP_113829.1 | 4 - 5 | 551 | 66 | |
XM_006242114.2 | XP_006242176.1 | 4 - 5 | 1343 | 66 | ||
GenBank mRNA | AF176343.1 | - | 1 - 2 | 141 | 66 | |
AF188699.1 | - | 4 - 5 | 551 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |