Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127600.1 | NP_001121072.1 | 1 - 2 | 261 | 60 | |
GenBank mRNA | BC161899.1 | - | 1 - 2 | 261 | 8367117 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |