Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108247.2 | NP_001101717.1 | 5 - 6 | 974 | 66 | |
XM_006256719.2 | XP_006256781.1 | 6 - 7 | 1515 | 66 | ||
XM_006256720.2 | XP_006256782.1 | 6 - 7 | 1772 | 66 | ||
XM_008773075.1 | XP_008771297.1 | 6 - 7 | 1743 | 66 | ||
GenBank mRNA | AF452730.1 | - | 6 - 7 | 1728 | 66 | |
BC167040.1 | - | 5 - 6 | 974 | 7135266 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |