Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131001.1 | NP_001124473.1 | 3 - 4 | 258 | 63 | |
GenBank mRNA | BC160864.1 | - | 3 - 4 | 258 | 7324585 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |