Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105982.1 | NP_001099452.1 | 4 - 5 | 712 | 77 | |
XM_006250373.2 | XP_006250435.1 | 4 - 5 | 866 | 77 | ||
XM_006250375.2 | XP_006250437.1 | 4 - 5 | 751 | 77 | ||
XM_008769820.1 | XP_008768042.1 | 4 - 5 | 866 | 77 | ||
XM_008769821.1 | XP_008768043.1 | 4 - 5 | 755 | 77 | ||
XM_008769822.1 | XP_008768044.1 | 4 - 5 | 844 | 77 | ||
XM_008769823.1 | XP_008768045.1 | 4 - 5 | 866 | 77 | ||
GenBank mRNA | FQ212123.1 | - | 4 - 5 | 696 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |