Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191935.1 | NP_001178864.1 | 2 - 3 | 695 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |