Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007667.2 | NP_001007668.1 | 5 - 6 | 632 | 72 | |
XM_008773228.1 | XP_008771450.1 | 5 - 6 | 1632 | 72 | ||
XM_008773229.1 | XP_008771451.1 | 5 - 6 | 1028 | 72 | ||
XM_008773230.1 | XP_008771452.1 | 6 - 7 | 667 | 72 | ||
GenBank mRNA | AY325208.1 | - | 497 | 72 | ||
BC060588.1 | - | 5 - 6 | 634 | 6921572 | 72 | |
FQ222039.1 | - | 5 - 6 | 514 | 72 | ||
FQ229984.1 | - | 5 - 6 | 516 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |