Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772932.1 | XP_008771154.1 | 11 - 12 | 1280 | 64 | |
XM_008772933.1 | XP_008771155.1 | 10 - 11 | 1142 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |