Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr25l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135894.1 | NP_001129366.1 | 4 - 5 | 1192 | 63 | |
XM_006240544.2 | XP_006240606.1 | 5 - 6 | 1409 | 63 | ||
XM_006240545.2 | XP_006240607.1 | 5 - 6 | 1254 | 63 | ||
XM_006240546.2 | XP_006240608.1 | 5 - 6 | 1272 | 63 | ||
XM_006240547.2 | XP_006240609.1 | 5 - 6 | 1480 | 63 | ||
GenBank mRNA | BC166712.1 | - | 4 - 5 | 1192 | 7930656 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |