Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198770.2 | NP_942065.1 | 2 - 3 | 297 | 76 | |
XM_006233597.2 | XP_006233659.1 | 295 | 76 | |||
GenBank mRNA | AJ488144.1 | - | 128 | 76 | ||
BC088093.1 | - | 2 - 3 | 297 | 7368043 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |