Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748666.3 | XP_003748714.1 | 16 - 17 | 1987 | 67 | |
XM_006227693.2 | XP_006227755.1 | 15 - 16 | 3643 | 67 | ||
XM_006227695.2 | XP_006227757.1 | 16 - 17 | 1701 | 67 | ||
XM_006227696.2 | XP_006227758.1 | 15 - 16 | 1620 | 67 | ||
XM_006227697.2 | XP_006227759.1 | 15 - 16 | 1906 | 67 | ||
XM_006227698.2 | XP_006227760.1 | 14 - 15 | 3568 | 67 | ||
XM_006227699.2 | XP_006227761.1 | 14 - 15 | 1466 | 67 | ||
XM_008758644.1 | XP_008756866.1 | 15 - 16 | 2822 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |