Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750582.3 | XP_003750630.1 | 6 - 7 | 835 | 76 | |
XM_006243865.2 | XP_006243927.1 | 6 - 7 | 835 | 76 | ||
XM_006243866.2 | XP_006243928.1 | 6 - 7 | 835 | 76 | ||
XM_006243867.2 | XP_006243929.1 | 6 - 7 | 835 | 76 | ||
XM_006243868.2 | XP_006243930.1 | 6 - 7 | 835 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |