Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013955.1 | NP_001013977.1 | 1 - 2 | 489 | 89 | |
XM_008766513.1 | XP_008764735.1 | 1 - 2 | 528 | 89 | ||
GenBank mRNA | BC087151.1 | - | 1 - 2 | 489 | 7131982 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |