Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126098.1 | NP_001119570.1 | 3 - 4 | 502 | 61 | |
XM_008765987.1 | XP_008764209.1 | 3 - 4 | 291 | 61 | ||
XM_008765988.1 | XP_008764210.1 | 3 - 4 | 328 | 61 | ||
XM_008765989.1 | XP_008764211.1 | 3 - 4 | 324 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |