Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109585.1 | NP_001103055.1 | 1 - 2 | 700 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |